
Ammonium chloride 2% (w/v) in aqueous solution
Catalog # RC62416
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:500 ml
- Cat. no.:RC62416
- Supplier No.:R0624000-500A
Specifications
Specification Test Results
- Appearance:Clear, colorless, odorless
Specification Test Results
About this item
:
:53.49 g/mol
Boiling Pt:100 °C
Melting Pt:0.0 °C
Density:1.0063 g/cm³
:GHS08
Storage Temperature:Ambient
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420