Orthophosphoric acid 0.1% (v/v) in aqueous solution
Catalog # RC5845-1
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:4 L
- Cat. no.:RC5845-1
- Supplier No.:R5845000-4A
Specifications
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 84):0.100 ± 0.002 % (v/v)
Specification Test Results
About this item
:
:98 g/mol
UN:1805
ADR:8,III
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340