ContactUsLinkComponent
Phosphate Buffer, for Cyanide, Ricca Chemical Company
Catalog # RC5810-32
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Phosphate Buffer, for Cyanide, Ricca Chemical Company
Catalog # RC5810-32
Supplier:  Ricca Chemical
Supplier Number:  R5810000-1A
Small Business Enterprise
CAS Number:  
Specifications
  • Size:
    32 oz.
  • Cat. no.:
    RC5810-32
  • Supplier No.:
    R5810000-1A
Specifications
About this item
Suitable for use in the following compendial methods: -ASTM D2036 D as Reagent 36.6 -APHA 4500-CN H as Reagent 4g