Order Entry
Canada
ContactUsLinkComponent
 
Mercury Standard, 1 mL = 0.1 mg Hg, 100 ppm Hg, Ricca Chemical Company
  RC4795-16
Small Business Enterprise
 :  
undefined
Mercury Standard, 1 mL = 0.1 mg Hg, 100 ppm Hg, Ricca Chemical Company
  RC4795-16
 :  R4795000-500A
Small Business Enterprise
 :  

 

  • Concentration:
    100 µg/ml
  • Matrix:
    1.3% HNO₃
  • Size:
    16 oz.
  • Cat. no.:
    RC4795-16
  • Element:
    Mercury
  • Supplier No.:
    R4795000-500A