Hydrochloric acid 6.25 N
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,II
:36.46
UN:1789
Specification Test Results
- Appearance:Clear, colorless to slight yellow-green, Hydrochloric odor
- Assay at 20 °C (traceable to NIST SRM 84):6.250 ± 0.005 N at 20.0 °C
Specification Test Results
Related Information
Frequently Bought Together