Hydrochloric acid 0.5167 N
RC3651-5
Small Business Enterprise
:
- Size:20 L
- Cat. no.:RC3651-5
- Supplier No.:R3651000-20F
- Appearance:Clear, colorless, slight Hydrochloric odor
- Assay at 20 °C (traceable to NIST SRM 723):0.5167 ± 0.0005 N at 20.0 °C
:
:8,II
:36.46
:1789
:7647-01-0
:HCl
:MFCD00011324