Order Entry
Canada
Orders LinkContactUsLinkComponent
Hydrochloric acid 0.1782 N

Hydrochloric acid 0.1782 N

Hydrochloric acid 0.1782 N
Supplier:  Ricca Chemical
Certificates
Hydrochloric acid 0.1782 N

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

1 mL = 5 mg CaO Lime

Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46
  • UN:1789
  • ADR:8,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear, colorless liquid
  • Assay at 20 °C (traceable to NIST SRM 723):
    0.1782 ± 0.0001 N at 20 °C
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 gal.
    Log in for availability
    Log in for availability
    Each (1US Gal)
    Each (1US Gal) - 221,33 $
    221,33 $
    Each (1US Gal)
       221,33 $

    Supplier Number: R3618000-4A
  • 5 gal.
    Log in for availability
    Log in for availability
    Each (5US Gal)
    Each (5US Gal) - 509,41 $
    509,41 $
    Each (5US Gal)
       509,41 $

    Supplier Number: R3618000-20F

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.