Order Entry
Canada
ContactUsLinkComponent
 
Hydrochloric acid 33.33% (v/v) aqueous solution (1 + 2), with 0.0005% (w/v) methyl orange indicator
Catalog # RC3582-1
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Hydrochloric acid 33.33% (v/v) aqueous solution (1 + 2), with 0.0005% (w/v) methyl orange indicator
Catalog # RC3582-1
Supplier:  Ricca Chemical
Supplier Number:  R3582000-4A
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    1 gal.
  • Cat. no.:
    RC3582-1
  • Supplier No.:
    R3582000-4A

Specifications

Specification Test Results

  • Appearance:
    Clear, orangish-red, Hydrochloric odor
  • Assay at 20 °C (traceable to NIST SRM 84):
    33.33 ± 0.08 % (v/v)

Specification Test Results

About this item

 :
ADR:8,II
 :36.46
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324