Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Acetone USP Test Solution (TS), buffered

1 L
Catalog # RC230-32
Supplier:   Ricca Chemical
 :  R0230000-1A
Acetone USP Test Solution (TS), buffered
Catalog # RC230-32
Supplier:  Ricca Chemical
CAS Number:  67-64-1
Acetone USP Test Solution (TS), buffered
Acetone USP Test Solution (TS), buffered

1 L

CAS Number:  67-64-1
Price
$225.24
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • UN:1090
  • ADR:3,II
Specifications

Specification Test Results

  • Appearance:
    Clear to slight haze, colorless, Acetone odor
  • pH at 25 °C (traceable to NIST SRM 186 & 191):
    5.9 ± 0.2 pH at 25.0 °C

Specifications

  • Size:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.