Calcium Standard, 1 ml = 0.2 mg CaCO₃, 200 ppm CaCO₃ (80.09 ppm Ca), Ricca Chemical Company
Catalog # RC179816
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:200 µg/ml CaCO₃
- Matrix:H₂O/tr. HCl
- Size:500 ml
- Cat. no.:RC179816
- Element:Calcium
- Supplier No.:R1798000-500A