96-Well Autosampler Vial Plate, with Inserts, MicroSolv™
Supplier: MicroSolv Technology Corp
Certificates
About this item
96 well plates with glass inserts.
- High grade, pure silicone rubber with PTFE coating
- Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass
- Chemically inert
The inserts are removable and the plates can be reused.
Frequently Bought Together