Vertical Exam Glove Dispenser, S-Curve Technologies
87000-005
- Color:Clear
- Material:Acrylic, ⅛
- Cat. no.:87000-005
- W×H×D:139.7×279.4×101.6 mm (5½×11×4")
- Supplier No.:EBD-1UP
Dispenser for a box of exam gloves-screws and wall anchors included.