Ammonium sulfate, crystals USP, ACS, FCC (GMP, ISO 9001, ISO22000)
89234-890
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Pail, red top, wide
- Size:12 kg
- Cat. no.:89234-890
- Supplier No.:50704000120
- Identification A & B:Corresponds to tests
- Assay (USP):99.0 - 100.5 %
- pH (USP):5 – 6
- Residue on ignition (USP):0.005 % Max.
- Insoluble matter (USP):0.005 % Max.
- Phosphate (USP):5 µg/g Max.
- Chloride (USP):5 ug/g Max.
- Nitrate (USP):0.001% Max.
- Iron (USP):5 µg/g Max
- Total aerobic microbial (USP/ACS):1000 cfu/g Max.
- Mold and yeasts (USP/ACS):10 cfu/g Max.
- Assay (FCC):99.0 – 100.5 %
- Appearance (FCC):Colorless or white crystals or granules that decompose at temperatures above 280°C.
- Solubility (FCC):One gram is soluble in about 1.5 ml of water. It is soluble in alcohol.
- Residue on ignition (FCC):0.25% Max.
- Lead (FCC):3 mg/kg Max.
- Selenium (FCC):0.003 % Max.
- Assay (ACS):99.0% Min.
- pH (ACS):5.0 – 6.0
- Residue on ignition (ACS):0.005 % Max.
- Insoluble matter (ACS):0.005 % Max.
- Phosphate (ACS):5 ppm Max.
- Chloride (ACS):5ppm Max.
- Nitrate (ACS):0.001% Max.
- Iron (ACS):5ppm Max.
- Total aerobic microbial:1000 cfu/g Max.
- Heavy metals (as Pb) (ACS):5 ppm Max.
- Bacterial endotoxins:2.5 EU/g Max.
:
:132.14 g/mol
:13,00559
:336…339 °C
:1.78 g/cm³ (20 °C)
:Ambient
:231-984-1
:01-2119455044-46
:7783-20-2
:(NH₄)₂SO₄
:MFCD00003391