ContactUsLinkComponent
LaMotte® AP Environmental Science Water Quality Assessment Module
LaMotte® AP Environmental Science Water Quality Assessment Module
Catalog # 470136-872
Supplier:  LaMotte
undefined
LaMotte® AP Environmental Science Water Quality Assessment Module
Catalog # 470136-872
Supplier:  LaMotte
Supplier Number:  5845
Specifications
  • Description:
    LaMotte AP Environmental Science Water Quality Assessment Module
  • Cat. no.:
    470136-872
  • Supplier No.:
    5845
  • No. of Parts:
    10 tests
Specifications
About this item
A Comprehensive Water Pollution Test Kit
  • Curriculum to teach assessment of water quality
  • Aligned to NGSS HS-LS2-1, HS-LS2-7, HS-ESS2-2, HS-ESS2-5, HS-ESS3-1, HS- ESS3-6
  • Operating Temperature 35 °C
This complete activity for your APES course is designed to satisfy water quality analysis requirements. It includes an extensive curriculum that uses the exploration of the Water Quality Index to teach students STEM-based skills that they will apply through classroom and field activities to satisfy Section VI (Water Pollution) of the AP Environmental Topics Outline.
Kit Contents: Ten nutrient - TTC/MacConkey BioPaddles for coliform testing; Salt/TDS/Temp Tracer for temperature and TDS determination; Teacher Resource CD-ROM containing Teacher and Student Guides, water quality test procedures, PowerPoint presentations, and Quick Time iPad/iPod videos for STEM design activities and STEM skills, Watershed Ecology, WQI, Point/Non-Point Pollution, glossary and free app; Free LaMotte BioPaddle colony; Identification app module for use with the Water Quality Educator.