Two Compartment Dispenser, Mid-Size, S-Curve Technologies
Supplier: S-Curve Technologies
Certificates
About this item
Mid-Size dispenser with 2 compartments and front access holes-only 9" off wall.
Designed for gloves, booties, bouffants, beard covers, etc.
Wall mount or free-standing, mounting bracket included.
Flame polished edges for smoothness.
Specifications
- Catalog No:
- 89208-694
- 82028-172
- PPE type:
- Multi-use
- Multi-use
- Material:
- PETG
- Acrylic
- No. of compartments:
- 2
- 2
- Dispenser type:
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable