Syringe Priming Kit, Hamilton Company
Catalog #: 89187-008
Supplier: Hamilton
Specifications
- Description:Syringe Priming Kit
- Cat. no.:89187-008
- Supplier No.:PRMKIT
Specifications
About this item
Hamilton Company offers the most comprehensive selection of standard and custom syringes, needles and accessories in the industry.
The syringe priming kit provides the necessary tools to ensure that the syringe and needle connection is completely primed. Trapped air can cause inaccurate dispenses and standard priming techniques are not sufficient when using capillary tubing. The glass capillary tubing traps a large volume of liquid that can only be overcome by using the priming kit. The Priming Kit contains one 250 μl Syringe, one 6 pack of 30 gauge needles, and one pack of septa.
Each is manufactured to achieve the highest level of accuracy and precision possible.
Ordering information: If you are looking for a syringe, needle point or accessory that is available but not listed here, please call Avantor to place a special order by phone.
Customers Who Bought This Also Bought