SoftGrip™ 8-Channel 50 µl Adjustable Volume Manual Pipettor Kit, Hamilton Company
Catalog #: 89050-812
Supplier: Hamilton
Specifications
- Volume:5–50 µL
- Description:SoftGrip™ Adjustable Volume Manual Pipettor Kit
- Cat. no.:89050-812
- No. of Channels:8
- Supplier No.:SSKIT-04
Specifications
About this item
The Hamilton line of pipettors puts a priority on addressing the issues of hand strain, fatigue, and other injuries.
- Features a dual indicator that improves the volume adjustment accuracy by removing parallax errors
- Slim ejection sleeve is made from a high strength, chemically resistant polymer
- Slim design allows the pipettor to reach the bottom of narrow tubes without the need for extended length tips
SoftGrip™ pipettors have a universal calibration key that reduces the need for costly calibration services. Calibration requires only minutes to perform and eliminates the downtime and cost of alternative calibration methods. The SoftGrip™ tip nozzle ensures a proper fit with universal pipettor tips. Unique color coding and labeling clearly indicates the working ranges of the adjustable volume pipettors.
Hamilton products set new standards for quality, comfort, and precision. The SoftGrip™ Adjustable volume manual pipettor kit includes an 8-channel, 50 µl SoftGrip™ Adjustable SoftGrip pipettor and pipettor stand.
Ordering information: For pipettor calibration labels, see 89050-826 series. For pipettor calibration key, see 89050-824 series.
Customers Who Bought This Also Bought