Potassium hydrogen carbonate 99.5-101.5% (dried basis), granular FCC, USP, Macron Fine Chemicals™
Catalog # CAMK673227
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: RegulatedCertificates
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Fiber drum
- Size:300 lbs.
- Cat. no.:CAMK673227
Specifications
Specification Test Results
- USP - Assay (KHCO₃) (dried basis):99.5 - 101.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Loss on Drying:≤0.25 %
- USP - Normal Carbonate:≤2.5 %
- FCC - Assay (KHCO₃) (dried basis):99.5 - 101.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Carbonate:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying:≤0.25 %
- Arsenic (As):≤3 ppm
- Solubility:Passes Test
- Appearance (white granules):Passes Test
Specification Test Results
About this item
:
:100.12 g/mol
Merck Index:13,07691
Melting Pt:292 °C
Density:2.17 g/cm³ (20 °C)
:Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:206-059-0
CAS Number:298-14-6
Formula:KHCO₃
MDL Number:MFCD00011402