Order Entry
Canada
Orders LinkContactUsLinkComponent
Urea ≥99% USP

Urea ≥99% USP

Urea ≥99% USP
MOC:  Regulated
Certificates
Urea ≥99% USP
MOC:  Regulated

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • GMP manufactured product
Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132.7 °C
  • Density:1.32 g/cm³ (20 °C)
  •  :Management of Change (MOC) category = R
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33

Documents

Specifications for Product Family

Specification Test Results

  • USP - Assay (NH₂CONH₂):
    98.0 - 102.0 %
  • USP - Alcohol Insoluble Matter:
    ≤ 0.04 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Organic impurities: Urea related compound A:
    ≤ 0.1 %
  • USP - Organic impurities: Any individual unspecified impurity:
    ≤ 0.1 %
  • USP - Organic impurities: Total impurities:
    ≤ 2.0 %
  • USP - Residue on Ignition:
    ≤ 0.1 %
  • Absorbance of a 5M Solution - 260 nm:
    ≤ 0.050
  • Absorbance of a 5M Solution - 280 nm:
    ≤ 0.030
  • Biuret:
    ≤ 0.01
  • Conductivity of 8.5M Solution at 25°C, (µmho/cm):
    ≤ 30
  • Cyanate (CNO):
    None Detected
  • Endotoxin Concentration (EU/g):
    ≤ 2.5
  • Insoluble Matter:
    ≤ 0.01 %
  • Water (H₂O):
    ≤ 0.5 %
  • Chloride (Cl):
    ≤ 1 ppm
  • Trace Impurities - Copper (Cu):
    ≤ 1 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 1 ppm
  • Trace Impurities - Lead (Pb):
    ≤ 5 ppm
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.