Order Entry
Canada
ContactUsLinkComponent
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Corrosive
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Catalog # CAJT3149-9
CAS Number:  
MOC:  RegulatedCertificates
undefined
Corrosive
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Catalog # CAJT3149-9
Supplier Number:  BAKR3149-09
CAS Number:  
MOC:  RegulatedCertificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    PE drum, black
  • Size:
    600 lbs.
  • Cat. no.:
    CAJT3149-9
  • Supplier No.:
    3149-09

Specifications

Specification Test Results

  • Assay (KOH):
    45.0 - 46.5 %
  • Color (APHA):
    ≤10
  • Appearance:
    Passes Test
  • Density (g/mL) at 25°C:
    1.45 - 1.47
  • Identification:
    Passes Test
  • Insoluble Matter:
    ≤30 ppm
  • Potassium Carbonate (K₂CO₃):
    ≤1000 ppm
  • Ammonium Hydroxide Precipitate:
    ≤30 ppm
  • Chloride (Cl):
    ≤30 ppm
  • Nitrogen Compounds (as N):
    ≤2 ppm
  • Phosphate (PO₄):
    ≤1 ppm
  • ACS - Sulfate (SO₄):
    ≤5 ppm
  • Trace Impurities - Aluminum (Al):
    ≤5.000 ppm
  • Trace Impurities - Chromium (Cr):
    ≤5.0 ppm
  • Trace Impurities - Copper (Cu):
    ≤0.050 ppm
  • Heavy Metals (as Ag):
    ≤3 ppm
  • Trace Impurities - Iron (Fe):
    ≤1 ppm
  • Trace Impurities - Lead (Pb):
    ≤2.000 ppm
  • Trace Impurities - Mercury (Hg):
    ≤0.10 ppm
  • Trace Impurities - Nickel (Ni):
    ≤1.000 ppm
  • Trace Impurities - Sodium (Na):
    ≤200.000 ppm
  • Trace Impurities - Zinc (Zn):
    ≤5.000 ppm

Specification Test Results

About this item

 :
 :56.11 g/mol
ADR:8,II
 :Management of Change (MOC) category = R
EINECS:215-181-3
UN:1814
 :GHS05
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553