

Sodium hydroxide 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
ADR:8,II
:Management of Change (MOC) category = R
UN:1824
:GHS05
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (NaOH):50 - 52 %
- Chloride (Cl):≤30 ppm
- Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤2 ppm
- Sodium Carbonate (Na₂CO₃):≤3.0 %
Specification Test Results
Related Information
Certificates
Search by Lot Number0339 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals Avantor Top Rated Guide
Product Literature
spec_JT0339-26 spec_JT0339-69 spec_JT0339-9
Product specification
Frequently Bought Together