Order Entry
Canada
ContactUsLinkComponent
Corning® Trypsin
Corning® Trypsin
Catalog # CA45000-666
Supplier:  Corning
CAS Number:  
undefined
Corning® Trypsin
Catalog # CA45000-666
Supplier:  Corning
Supplier Number:  25-054-CI
CAS Number:  

Specifications

  • Size:
    100 ml
  • Storage conditions:
    Frozen
  • Shipping conditions:
    Frozen
  • Enzyme name:
    Trypsin
  • Formulation:
    Trypsin 10X, 2.5% Trypsin in HBSS without Calcium, Magnesium, and Phenol Red
  • Cat. no.:
    CA45000-666
  • Supplier No.:
    25-054-CI

Specifications

About this item

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging

  • Trypsin 10X 2,5% Trypsin in HBSS without calcium, magnesium, and phenol red Porcine Parvovirus Tested

Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin.