
Sodium carbonate, anhydrous 99.5-100.5%, EMPROVE® EXPERT Ph. Eur., BP suitable for use as excipient, SAFC®
Catalog # CA1.06398.5000
Supplier: EMD MILLIPORE # PCS CA EXCEP
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:5 kg
- Cat. no.:CA1.06398.5000
- Supplier No.:1.06398.5000
Specifications
Specification Test Results
- Assay (acidimetric; calculated on dried substance):99.5-100.5%
- Alkalihydroxides and alkalihydrogen-carbonates:To pass test
- Appearance of solution:To pass test
- Arsenic (As):0.0005% max
- Chloride (Cl):0.0125% max
- Heavy metals (as Pb):0.001% max
- Identity test A:To pass test
- Identity test B:To pass test
- Iron (Fe):0.005% max
- Loss on drying (300°C):0.5% max
- Residual Solvents (Ph.Eur./USP/ICH):Excluded by manufacturing process
- Sulfate (SO4):0.025% max
Specification Test Results
About this item
:
:105.99 g/mol
Merck Index:14,08596
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494