Orders LinkContactUsLinkComponent
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Supplier:   Corning
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Supplier:  Corning
Certificates
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
13 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Minimum Essential Medium (Mod.)(EMEM) based on Basal Medium Eagle (BME) formulation to aid cell propagation.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. This formulation contains Earle's salts and with or without L-glutamine.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CA45000-382
  • CA45000-384
  • CA45000-386
  • CA45000-388
  • CA45000-394
  • CA45000-582
  • CA45000-584
  • CA45000-586
  • CA45000-592
  • CA45000-594
  • CA45000-596
  • CA45000-380
  • CA11028-536
  • Storage temperature:
  • Refrigerate
  • Refrigerate
  • Refrigerate
  • Refrigerate
  • Refrigerate
  • Refrigerate
  • Refrigerate
  • Refrigerate
  • Refrigerate
  • Refrigerate
  • Refrigerate
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • CA45000-382
    Minimum Essential Medium (MEM) with Earle's salts and L-glutamine
    Minimum Essential Medium (MEM) with Earle's salts and L-glutamine
    Log in for availability
    Log in for availability
    Pack of 6 (1l)
    Pack of 6 (1l) - $269.00
    $269.00
    Pack of 6 (1l)
       $269.00
    Supplier Number: 10-010-CM
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-382
    Storage temperature
    Refrigerate
  • CA45000-384
    Minimum Essential Medium (MEM) with Earle's salts, without L-glutamine
    Minimum Essential Medium (MEM) with Earle's salts, without L-glutamine
    Log in for availability
    Log in for availability
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $160.00
    $160.00
    Pack of 6 (500ml)
       $160.00
    Supplier Number: 15-010-CV
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-384
    Storage temperature
    Refrigerate
  • CA45000-386
    Minimum Essential Medium (MEM) with Earle's salts, without L-glutamine
    Minimum Essential Medium (MEM) with Earle's salts, without L-glutamine
    Log in for availability
    Log in for availability
    Pack of 6 (1l)
    Pack of 6 (1l) - $314.00
    $314.00
    Pack of 6 (1l)
       $314.00
    Supplier Number: 15-010-CM
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-386
    Storage temperature
    Refrigerate
  • CA45000-388
    Minimum Essential Medium (MEM) with Earle's salts, without L-glutamine and phenol red
    Minimum Essential Medium (MEM) with Earle's salts, without L-glutamine and phenol red
    Log in for availability
    Log in for availability
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $217.00
    $217.00
    Pack of 6 (500ml)
       $217.00
    Supplier Number: 17-305-CV
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-388
    Storage temperature
    Refrigerate
  • CA45000-394
    Minimum Essential Medium (MEM) (For Suspension Cultures) without L-glutamine, calcium, and magnesium
    Minimum Essential Medium (MEM) (For Suspension Cultures) without L-glutamine, calcium, and magnesium
    Log in for availability
    Log in for availability
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $559.00
    $559.00
    Pack of 6 (500ml)
       $559.00
    Supplier Number: 15-015-CV
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-394
    Storage temperature
    Refrigerate
  • CA45000-582
    Minimum Essential Medium (MEM), Powder with Earle's salts and L-glutamine, without sodium bicarbonate
    Minimum Essential Medium (MEM), Powder with Earle's salts and L-glutamine, without sodium bicarbonate
    Log in for availability
    Log in for availability
    Each (10l)
    Each (10l) - $58.50
    $58.50
    Each (10l)
       $58.50
    Supplier Number: 50-010-PB
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-582
    Storage temperature
    Refrigerate
  • CA45000-584
    Minimum Essential Medium (MEM), Powder with Earle's salts and L-glutamine, without sodium bicarbonate
    Minimum Essential Medium (MEM), Powder with Earle's salts and L-glutamine, without sodium bicarbonate
    Log in for availability
    Log in for availability
    Each (50l)
    Each (50l) - $194.00
    $194.00
    Each (50l)
       $194.00
    Supplier Number: 50-010-PC
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-584
    Storage temperature
    Refrigerate
  • CA45000-586
    Minimum Essential Medium (MEM), powder with Earle's salts, without L-glutamine, phenol red, and sodium bicarbonate
    Minimum Essential Medium (MEM), powder with Earle's salts, without L-glutamine, phenol red, and sodium bicarbonate
    Log in for availability
    Log in for availability
    Each (10l)
    Each (10l) - $74.80
    $74.80
    Each (10l)
       $74.80
    Supplier Number: 90-009-PB
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-586
    Storage temperature
  • CA45000-592
    Minimum Essential Medium (MEM), Powder (Autoclavable) with Earle's salts, without L-glutamine and sodium bicarbonate
    Minimum Essential Medium (MEM), Powder (Autoclavable) with Earle's salts, without L-glutamine and sodium bicarbonate
    Log in for availability
    Log in for availability
    Each (50l)
    Each (50l) - $105.00
    $105.00
    Each (50l)
       $105.00
    Supplier Number: 51-010-PC
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-592
    Storage temperature
    Refrigerate
  • CA45000-594
    Minimum Essential Medium (MEM), Powder with Earle's salts, L-glutamine, and nonesstential amino acids without sodium bicarbonate
    Minimum Essential Medium (MEM), Powder with Earle's salts, L-glutamine, and nonesstential amino acids without sodium bicarbonate
    Log in for availability
    Log in for availability
    Each (10l)
    Each (10l) - $58.00
    $58.00
    Each (10l)
       $58.00
    Supplier Number: 50-011-PB
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-594
    Storage temperature
    Refrigerate
  • CA45000-596
    Minimum Essential Medium (MEM), Powder with Earle's salts, L-glutamine, and nonesstential amino acids without sodium bicarbonate
    Minimum Essential Medium (MEM), Powder with Earle's salts, L-glutamine, and nonesstential amino acids without sodium bicarbonate
    Log in for availability
    Log in for availability
    Each (50l)
    Each (50l) - $86.00
    $86.00
    Each (50l)
       $86.00
    Supplier Number: 50-011-PC
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-596
    Storage temperature
    Refrigerate
  • CA45000-380
    Minimum Essential Medium (MEM) with Earle's salts and L-glutamine
    Minimum Essential Medium (MEM) with Earle's salts and L-glutamine
    Log in for availability
    Log in for availability
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $173.00
    $173.00
    Pack of 6 (500ml)
       $173.00
    Supplier Number: 10-010-CV
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-380
    Storage temperature
    Refrigerate
  • CA11028-536
    Minimum Essential Medium (MEM), Sodium bicarbonate, NEAA, L-glutamine, sodium pyruvate
    Minimum Essential Medium (MEM), Sodium bicarbonate, NEAA, L-glutamine, sodium pyruvate
    Log in for availability
    Log in for availability
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $241.00
    $241.00
    Pack of 6 (500ml)
       $241.00
    Supplier Number: 10-009-CV
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA11028-536
    Storage temperature