Order Entry
Canada
Orders LinkContactUsLinkComponent
Corning® MEM Vitamins 100X Solution, Corning

Corning® MEM Vitamins 100X Solution, Corning

MEM Vitamins 100X Solution, 100 ml
Catalog # CA45000-702
Supplier:   Corning
Supplier #:  25-020-CI
Corning® MEM Vitamins 100X Solution, Corning
Catalog # CA45000-702
Supplier:  Corning
CAS Number:  
Corning® MEM Vitamins 100X Solution, Corning
Corning® MEM Vitamins 100X Solution, Corning

MEM Vitamins 100X Solution, 100 ml

CAS Number:  
Price
$179.00
Log in for availability
Pack of 6 (100ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The addition of MEM vitamins to medium can improve growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells

This concentrated supplement allows for efficient addition to media with no risk of dilution of media components.

Specifications

Specifications

  • Description:
    MEM Vitamins 100X Solution
  • Volume:
    100 ml
  • Cat. No.:
    CA45000-702
  • Storage temperature:
    Frozen
  • Shipping temperature:
    Frozen

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.