Corning® Medium 199 (Mod.), Corning
About this item
Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
It was originally developed as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation.
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
It was originally developed as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation.
Documents
Certificates
Search by Lot Number3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture.pdf
Product Literature
Cell Culture Media, Molecular Buffers and Reagents
Additional Product Details
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA45000-378
- CA45000-570
- CA45000-574
- Storage temperature:
- 2...8 °C
- 2...8 °C
- 2...8 °C
Specifications
Product Family Options
Product Information
- Media FormatAvailabilityPrice
- CA45000-378Medium 199 (Mod.) with Earle's salts and L-glutamineMedium 199 (Mod.) with Earle's salts and L-glutamineLiquidLog in for availabilityPack of 6 (500ml)$189.00Supplier Number: 10-060-CVItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.CA45000-378Storage temperature2...8 °C - CA45000-570Medium 199 (Mod.), Powder, with Earle's salts and L-glutamine without sodium bicarbonateMedium 199 (Mod.), Powder, with Earle's salts and L-glutamine without sodium bicarbonatePowderLog in for availabilityEach (10l)$133.00Supplier Number: 50-050-PBItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.CA45000-570Storage temperature2...8 °C - CA45000-574Medium 199 (Mod.), Powder [+] Earle's salts [-] sodium bicarbonate, L-glutamine, phenol redMedium 199 (Mod.), Powder [+] Earle's salts [-] sodium bicarbonate, L-glutamine, phenol redPowderLog in for availabilityEach (10l)$138.00Supplier Number: 90-050-PBItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.CA45000-574Storage temperature2...8 °C