Corning® Leibovitz's L-15 with L-glutamine, Corning
CA45000-372
: Corning
:
- Description:Leibovitz's L-15 with L-glutamine
- Storage temperature:Refrigerate
- Shipping temperature:Ambient
- Cat. no.:CA45000-372
- Volume:500 ml
- Supplier No.:10-045-CV
Leibovitz's L15 medium was originally developed without a sodium bicaronate buffer system for use in non-CO2 equilibrated environments
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
It is buffered by phosphates and the amino acid L-arginine, along with L-histidine and L-cysteine. The formation of acid metabolite byproducts is prevented by replacing glucose with galactose and sodium pyruvate. Examples of usage include the propagation of HEP-2, monkey kidney cells and primary explants of embryonic and adult human tissue. Leibovitz's L-15 supports established cell lines as well as primary explants of embryonic and adult human tissue.