Corning® MEM (Minimum Essential Medium), Improved MEM (Richter's Mod.), Corning
: Corning
A modified forumulation of Minimal Essential Medium, Improved Minimum Essential Medium (IMEM) contains a higher concentration of many components used in MEM along with additional amino acids, vitamins, and metabolites.
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Formulated with cell culture grade WFI water
- Manufactured in a cGMP environment"
- Storage temperature:Refrigerate
Frequently Bought Together