Corning® HT (Hypoxanthine, Thymidine), Corning
Catalog # CA45000-688
Supplier: Corning
CAS Number:
Specifications
- Description:HT (Hypoxanthine, Thymidine)
- Size:100 ml
- Shipping conditions:Frozen
- Storage temperature:Frozen
- Cat. no.:CA45000-688
- Supplier No.:25-047-CI
Specifications
About this item
HT media is a 'rescue media' that provides pre-formed purines and pyrimidines to overcome the effects of residual intracellular aminopterin.
- Complete Certificate of Analysis available for each production lot, along with full formulation information
- Produced under the highest industry standards to ensure superior results