Order Entry
Canada
ContactUsLinkComponent
 
Corning® F-12K Nutrient Mixture (Kaighn's Mod.) with L-glutamine, Corning
Catalog # CA45000-354
Supplier:  Corning
CAS Number:  
undefined
Corning® F-12K Nutrient Mixture (Kaighn's Mod.) with L-glutamine, Corning
Catalog # CA45000-354
Supplier:  Corning
Supplier Number:  10-025-CV
CAS Number:  

Specifications

  • Description:
    F-12K Nutrient Mixture (Kaighn's Mod.) with L-glutamine
  • Storage temperature:
    Refrigerate
  • Cat. no.:
    CA45000-354
  • Supplier No.:
    10-025-CV

Specifications

About this item

A modification of Ham's F-12, F-12K contains higher concentrations of amino acids, sodium pyruvate and other components than the original

  • Low endotoxin: >0.25EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

It is formulated to support the growth of cells including differentiated rat and chicken cell types and primary human hepatocytes.