Specifications
- Description:Corning® Cellstripper® Liquid
- Storage temperature:Ambient
- Shipping temperature:Ambient
- Cat. no.:CA45000-668
- Volume:100 mL
- Supplier No.:25-056-CI
Specifications
About this item
Cellstripper® is a non-enzymatic cell dissociation solution formulated with a proprietary mixture of chelators that gently dislodge adherent cells in tissue culture. It is an alternative to trypsin when used with either serum-free or serum-supplemented media. Because Cellstripper® is a gentler reagent than trypsin, cells can be exposed to Cellstripper for longer periods of time. Cellstripper® achieves superior results in comparison to enzymatic dissociation products.