Syringe, Chromatography, LC Injection, Economy 6-Pack, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Basic 10 microliter LC injection syringe with epoxy cemented 304 stainless steel needles permanently attached, or with removable needles
Needles are 26 gauge.
Ordering information: Supplied in convenient package of six. Includes convenient storage container.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.