Midget Impinger, Ace-Threds, Sintered Glass Filter, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This piece is used for sampling air volumes at low jet velocity
Bottle joint is reinforced [ST] 24/40 outer. Tube uses #7 “Ace-Safe” removable hose connections for easy connect/disconnect. Nozzle has sintered glass filter, Porosity A (145-175 microns) or Porosity B (70-100 micron). Bottle has encircling graduations from 0 to 25mL or 0 to 30mL in 5mL divisions. Fabricated from borosilicate glass.
Ordering information: Complete item consists of bottle, tube, connectors and bushings. Items can also be purchased separately.
Frequently Bought Together