About this item
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 21,250 cm2 polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
Specifications
- Catalog No:
- 89090-254
- Environmentally Preferable:
- Low Manufacturing Impact
- Media volume:
- 1500 ml
- Surface:
- Tissue Culture Treated
- Environmental certification:
- RECs, ISO 14001 - Kennebunk