Orders LinkContactUsLinkComponent
Visual Scientifics Projectile Launcher
Visual scientifics projectile launcher
Catalog # 470135-574
Supplier:   Wards
 :  PTPROJ
Visual Scientifics Projectile Launcher
Catalog # 470135-574
Supplier:  Wards
Visual Scientifics Projectile Launcher
Visual Scientifics Projectile Launcher
Visual scientifics projectile launcher
Price
$160.00
Log in for availability
Each
Product Details & Documents
This inquiry-based activity allows students to investigate and verify projectile equations.
  • 23 page lab manual included, with activities and extensions
  • Platform neutral - works with your preferred probeware/video analysis tools
Students explore two-dimensional projectile motion. Using video analysis with two extension activities, students verify theoretical modeling and use experimental results to meet design criteria. The included lab manual provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and Common Core concepts.
 : Meets 12 Next Generation Science Standards concepts and 10 common core standards.
 : Photogate system required, but sold separately (470317-428 recommended). Visual Scientifics Base (470148-822), required, but sold separately.
 : This kit includes a projectile launcher, mounting hardware, protractor with bob, and ball. Recommended accessories: video camera, graphical analysis software, carbon paper, and masking tape.
Specifications
  • Description:
    Visual scientifics projectile launcher
Accessories
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Visual scientifics base
    Visual scientifics base
    470148-822
    25×16×2.5"
    Log in for availability
    Each - $214.85