Orders LinkContactUsLinkComponent
Visual Scientifics Free Fall
Visual Scientifics Free Fall
Visual scientifics free fall
Catalog # 470135-572
Supplier:   Wards
Supplier #:  PTFFAL
Visual Scientifics Free Fall
Catalog # 470135-572
Supplier:  Wards
Visual Scientifics Free Fall
Visual Scientifics Free Fall
Visual scientifics free fall
Price
$151.50
Log in for availability
Each
Product Details & Documents
Students measure acceleration due to gravity in this free-fall activity. Video analysis allows modeling of air resistance.
  • 20 page lab manual included, with activities and extensions
  • Covers 12 NGSS and 13 common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
This free fall system makes it easy to study acceleration due to gravity, and video analysis extension activities make it possible for students to model air resistance. The included lab manual provides approximately three hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and common core concepts.
Ordering information: Photogate system required, but sold separately (470317-428 recommended). Visual Scientifics Base (470148-822), required, but sold separately.
Delivery information: This kit includes a release mechanism, steel ball, and landing pad. A photogate system is required, but not included. Recommended: video camera and graphical analysis software.
Specifications
  • Description:
    Visual scientifics free fall
Accessories
Product Information
  • Catalog #
    Size
    Availability
    Price
  • Visual scientifics base
    Visual scientifics base
    470148-822
    25×16×2.5"
    Log in for availability
    Each - $214.85
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Fundamentals of physics photogate system
    Fundamentals of physics photogate system
    470014-466
    16×10×6 cm
    0.5 kg (1.1 lbs.)
    Log in for availability
    Each - $530.35