Specifications
- Cat. no.:CA26108-10
- Supplier No.:26108-10
Specifications
About this item
Hach Paddle Testers allow fast, easy screening of water samples, solid surfaces, and nonviscous liquids where unsanitary conditions may exist
The paddle is a double-sided slide attached to the vial cap. Each side of the slide is used to perform a separate test. The slide has a molded-in grid for easy colony counting without opening the optically clear, leakproof vial. Paddle Tester, Total Aerobic Bacteria/Yeast and Mold (Tryptone Glucose Extract Agar/Sabouraud Dextrose Agar), pk/10.