ContactUsLinkComponent
Phosphorus, Orthophosphate (reactive) Test Strips, Hach
Phosphorus, Orthophosphate (reactive) Test Strips, Hach
Catalog # CA11014-410
Supplier:  Hach
CAS Number:  
Phosphorus, Orthophosphate (reactive) Test Strips, Hach
Catalog # CA11014-410
Supplier:  Hach
Supplier Number:  2757150
CAS Number:  
Specifications
  • Cat. no.:
    CA11014-410
  • Range:
    0 - 50 mg/L
  • Supplier No.:
    2757150
  • No. of Tests:
    50
Specifications
About this item
Hach Test Strips are easy to use and read, disposable, and inexpensive
In the field, technicians can test many samples in just minutes and make immediate evaluations on-site. In the lab, Hach test strips are ideal in pre-test screening to detect the presence of materials that might interfere with lab testing.