Spectra/Por® Micro Float-A-Lyzer Dialysis Devices, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Certificates
About this item
Spectrum introduces the new Micro Float-A-Lyzer® to expand our popular line of ready-to-use dialysis devices for the easy and efficient dialysis of very small sample volumes (100 to 500 µl).
Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) tubular membrane into an easy-to-use design making laboroatory microdialysis easier than ever. Micro Float-A-Lyzer is available in 2 volumes sizes: 100 to 200 µl and 400 to 500 µl; and 7 concise MWCO's ranging from 100 to 100000 Daltons.
Ordering information: 1 ml syringe included.
Customers Who Bought This Also Bought