Filter Disc, Packing Support, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This filter disc is for use with column adapters.
Available in polyethylene (100 µm pore size), and borosilicate glass. The glass discs feature four different porosities: Porosity A (145 to 174 µm), Porosity B (70 to 100 µm), Porosity C (25 to 50 µm) and Porosity D (10 to 20 µm).
Ordering information: Caution: These discs are intended to be removable. However, because of the tight fit, the glass disc may break when being removed.
Frequently Bought Together