ContactUsLinkComponent
5-in-1 Water Quality Test Strips, Hach
5-in-1 Water Quality Test Strips, Hach
  CA2755250
 :  Hach
 :  
5-in-1 Water Quality Test Strips, Hach
  CA2755250
 :  Hach
 :  2755250
 :  
  • Cat. no.:
    CA2755250
  • Range:
    Various
  • Supplier No.:
    2755250
  • No. of Tests:
    50
5-in-1 test strips are multi-parameter test strips test offering five water quality measurements on 1 strip.
  • Free Chlorine (010 mg/L)
  • Total Chlorine (010 mg/L)
  • Total Hardness (as CaCO3, 025 gpg and 0425 mg/L)
  • Total Alkalinity (as CaCO3, 0240 mg/L)
  • pH (6.2 8.4)
With Hach Water Quality Test Strips, technicians in the field can test many samples in only a few minutes and make immediate evaluations on-site. No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing.