ContactUsLinkComponent
Guardian™ Airflow Monitor, Labconco®
Guardian™ Airflow Monitor, Labconco®
Catalog # 89233-240
Supplier:  Labconco
Product available on GSA Advantage®
undefined
Guardian™ Airflow Monitor, Labconco®
Catalog # 89233-240
Supplier:  Labconco
Supplier Number:  9413301
Product available on GSA Advantage®
Specifications
  • Description:
    Guardian™ Airflow Monitor, 230V, 50/60Hz
  • Environmentally Preferable:
  • Cat. no.:
    89233-240
  • REDISHIP:
    No
  • Supplier No.:
    9413301
Specifications
About this item
Guardian Airflow Monitors are designed to continuously monitor airflow through Labconco fume hoods
Provides visual and audible alarms for abnormal airflow conditions.