ContactUsLinkComponent
SP Bel-Art Test Tube Capper, Bel-Art Products, a part of SP
SP Bel-Art Test Tube Capper, Bel-Art Products, a part of SP
Catalog # 89233-990
undefined
SP Bel-Art Test Tube Capper, Bel-Art Products, a part of SP
Catalog # 89233-990
Supplier Number:  F18799-0000
Specifications
  • Description:
    Test Tube Capper, 44×89 m m (1¾ × 3½”)
  • Cat. no.:
    89233-990
  • Supplier No.:
    F187990000
Specifications
About this item
The vinyl-covered handhold fits comfortably in the palm of the hand while the polypropylene tip presses down on the cap to securely position it within the test tube.
  • Vinyl-covered handhold
  • Polypropylene tip secures cap in position
  • Reduced finger soreness and hand stress
Capping can be performed on glass or plastic test tubes in a rack, or while being held in the hand
With the Scienceware® Test Tube Capper, you can use your entire hand for leverage, reducing finger soreness and hand stress.
It is designed for use with recessed center test tube caps.