About this item
HyClone Bovine Calf Serum, U.S, can be used as an effective alternative to foetal bovine serum (FBS).
- Filtered through sequential 100 nm pore size rated filters
- Extensive quality, performance and analytical testing
- Endotoxin Concentration ≤ 10 EU/mL
- Storage Temperature is –10 °C
Origin is carefully collected, processed and filtered in U.S. abbatoirs. Ensures excellent culture results without safety concerns.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA16777-206
- CA82007-424
- CA76534-652
- CA76237-690
- CA76237-694
- CA76534-626
- CA76324-872
- CA76237-688
- CA16777-204
- Origin:
- US
- US
- US
- US
- US
- US
- US
- US
- US
- Endotoxin:
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- Hemoglobin:
- <30 mg/dl
- <30 mg/dl
- <30 mg/dl
- <30 mg/dl
- <30 mg/dl
- <30 mg/dl
- <30 mg/dl
- <30 mg/dl
- <30 mg/dl
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
HyClone™ Cell Culture Serum products