Flask, Nephelo Culture, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This Nephelo culture flask has a long-style neck that accommodates a 25mm stainless steel or plastic closure
Ideal for growing, and mixing of aerobic and anaerobic cultures. Side arm fits standard spectrophotometers. Fabricated from borosilicate glass.
Ordering information: Closure NOT included.
Frequently Bought Together