Starch 2% in aqueous solution stabilized for iodometric titrations mercury-free
Catalog # RC805516
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly
- Size:500 ml
- Cat. no.:RC805516
- Supplier No.:R8055000-500A
Specifications
Specification Test Results
- Appearance:Translucent, odorless
- Suitability for Use:Colorless (Iodine absent) - Blue (Iodine present)
Specification Test Results
About this item
:
:162.14 g/mol
Storage Temperature:Ambient
EINECS:232-679-6
CAS Number:9005-25-8
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026