Titanium ICP Standard, 1 ml = 10 mg Ti (10,000 ppm Ti)(NH₄)2TiF₆ in H₂O / trace HF, Ricca Chemical Company
Catalog # RCPTI10KW-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:10000 µg/ml
- Matrix:H₂O/tr. HF
- Pack type:Poly
- Size:100 ml
- Cat. no.:RCPTI10KW-100
- Element:Titanium
- Supplier No.:RPTI10KW-100N
Specifications
About this item
10,000 ppm = 10000 µg/mL