Order Entry
Canada
ContactUsLinkComponent
Tantalum AA Standard, 1 ml = 1 mg Ta (1000 ppm Ta)(NH₄)₂TaF₇ in H₂O / trace HF, Ricca Chemical Company
Tantalum AA Standard, 1 ml = 1 mg Ta (1000 ppm Ta)(NH₄)₂TaF₇ in H₂O / trace HF, Ricca Chemical Company
Catalog # RCATA1KW-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Tantalum AA Standard, 1 ml = 1 mg Ta (1000 ppm Ta)(NH₄)₂TaF₇ in H₂O / trace HF, Ricca Chemical Company
Catalog # RCATA1KW-100
Supplier:  Ricca Chemical
Supplier Number:  RATA1KW0-100N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. no.:
    RCATA1KW-100
  • Element:
    Tantalum
  • Supplier No.:
    RATA1KW0-100N

Specifications

About this item