Order Entry
Canada
ContactUsLinkComponent
Selenium ICP Standard, 1 ml = 10 mg Se (10,000 ppm Se)Se in 5% HNO₃, Ricca Chemical Company
Selenium ICP Standard, 1 ml = 10 mg Se (10,000 ppm Se)Se in 5% HNO₃, Ricca Chemical Company
Catalog # RCPSE10KN-50
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Selenium ICP Standard, 1 ml = 10 mg Se (10,000 ppm Se)Se in 5% HNO₃, Ricca Chemical Company
Catalog # RCPSE10KN-50
Supplier:  Ricca Chemical
Supplier Number:  RPSE10KN-50N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    10000 µg/ml
  • Matrix:
    5% HNO₃
  • Pack type:
    Poly
  • Size:
    50 ml
  • Cat. no.:
    RCPSE10KN-50
  • Element:
    Selenium
  • Supplier No.:
    RPSE10KN-50N

Specifications

About this item