Order Entry
Canada
ContactUsLinkComponent
Scandium ICP Standard, 1 ml = 10 mg Sc (10,000 ppm Sc)Sc₂O₃ in 5% HNO₃, Ricca Chemical Company
Scandium ICP Standard, 1 ml = 10 mg Sc (10,000 ppm Sc)Sc₂O₃ in 5% HNO₃, Ricca Chemical Company
Catalog # RCPSC10KN-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Scandium ICP Standard, 1 ml = 10 mg Sc (10,000 ppm Sc)Sc₂O₃ in 5% HNO₃, Ricca Chemical Company
Catalog # RCPSC10KN-100
Supplier:  Ricca Chemical
Supplier Number:  RPSC10KN-100N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    10000 µg/ml
  • Matrix:
    5% HNO₃
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. no.:
    RCPSC10KN-100
  • Element:
    Scandium
  • Supplier No.:
    RPSC10KN-100N

Specifications

About this item